MAL-PEG8-NHS ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Mal-amido-PEG8-NHS, MAL-dPEG®8-NHS ester, Maleimide-PEG8-succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22,25,28-octaoxa-4-azahentriacontan-31-oate
BIPG1480-100MG
BIPG1480-500MG
BIPG1480-1G
APOSBIPG1480-100MGEA
6220
CZK
APOSBIPG1480-100MG
APOSBIPG1480-500MG
APOSBIPG1480-1G
MAL-PEG8-NHS ester
MAL-PEG8-NHS ester
Vzorec:
C₃₀H₄₇ N₃O₁₅ MW: 689.71 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11041144 CAS číslo: 756525-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...