Tisk…

Mal-amido-PEG6-NHS

Dodavatel: Apollo Scientific

Synonyms: 4,7,10,13,16,19-Hexaoxa-22-azapentacosanoic acid, 25-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-23-oxo-, 2,5-dioxo-1-pyrrolidinyl ester, PEGylated, Maleimide-NH-PEG6-CH2CH2COONHS ester

BIPG1479-100MG BIPG1479-250MG
APOSBIPG1479-100MGEA 1990 CZK
APOSBIPG1479-100MG APOSBIPG1479-250MG
Mal-amido-PEG6-NHS
Mal-amido-PEG6-NHS
Vzorec: C₂₆H₃₉N₃O₁₃
MW: 601,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11041093
CAS číslo: 1137109-21-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR