Tisk…

Mal-amido-PEG4-NHS

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,5-Dioxopyrrolidin-1-yl 19-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-17-oxo-4,7,10,13-tetraoxa-16-azanonadecan-1-oate, Mal-amido-PEG4-NHS ester, Maleimide-PEG4-succinimidyl Ester, Maleimide-PEG4-NHS Ester

BIPG1478-50MG BIPG1478-100MG BIPG1478-250MG
APOSBIPG1478-50MGEA 2900 CZK
APOSBIPG1478-50MG APOSBIPG1478-100MG APOSBIPG1478-250MG
Mal-amido-PEG4-NHS
Mal-amido-PEG4-NHS
Vzorec: C₂₂H₃₁N₃O₁₁
Storage Temperature: Freezer
MDL Number: MFCD11041113
CAS číslo: 756525-99-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR