Mal-amido-PEG12-acid
Dodavatel: Apollo Scientific
Synonyms:
Mal-amido-PEG12-acid
BIPG1476-100MG
BIPG1476-50MG
BIPG1476-250MG
APOSBIPG1476-100MGEA
5790
CZK
APOSBIPG1476-100MG
APOSBIPG1476-50MG
APOSBIPG1476-250MG
Mal-amido-PEG12-acid
Mal-amido-PEG12-acid
Vzorec:
C₃₄H₆₀N₂O₁₇ Storage Temperature: Ambient |
MDL Number:
MFCD13184984 CAS číslo: 871133-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...