Fmoc-N-amido-dPEG8-acid
Dodavatel: Apollo Scientific
|
Varování
|
BIPG1402-100MG
BIPG1402-500MG
APOSBIPG1402-100MGEA
7670
CZK
APOSBIPG1402-100MG
APOSBIPG1402-500MG
Fmoc-N-amido-dPEG8-acid
Fmoc-N-amido-dPEG8-acid
Vzorec:
C₃₄H₄₉NO₁₂ Storage Temperature: Freezer |
CAS číslo:
756526-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...