Tisk…

(Fmoc-amino)-PEG6-carboxylic acid

Dodavatel: Apollo Scientific

BIPG1400-100MG BIPG1400-500MG BIPG1400-1G
APOSBIPG1400-100MGEA 3060 CZK
APOSBIPG1400-100MG APOSBIPG1400-500MG APOSBIPG1400-1G
(Fmoc-amino)-PEG6-carboxylic acid
(Fmoc-amino)-PEG6-carboxylic acid
Vzorec: C₃₀H₄₁NO₁₀
Storage Temperature: Ambient
MDL Number: MFCD06656474
CAS číslo: 882847-34-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR