Tisk…

Fmoc-N-amido-dPEG®5-acid

Dodavatel: Apollo Scientific

BIPG1399-1G
APOSBIPG1399-1GEA 2530 CZK
APOSBIPG1399-1G
Fmoc-N-amido-dPEG®5-acid
Fmoc-N-amido-dPEG5-acid
Vzorec: C₂₈H₃₇NO₉
MW: 531,59 g/mol
Storage Temperature: Ambient
CAS číslo: 882847-32-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR