Tisk…

Fmoc-N-amido-dPEG®3-acid

Dodavatel: Apollo Scientific

BIPG1397-500MG BIPG1397-1G BIPG1397-100MG
APOSBIPG1397-500MGEA 1450 CZK
APOSBIPG1397-500MG APOSBIPG1397-1G APOSBIPG1397-100MG
Fmoc-N-amido-dPEG®3-acid
Fmoc-N-amido-dPEG3-acid
Vzorec: C₂₄H₂₉NO₇
MW: 443,49 g/mol
Storage Temperature: Ambient
CAS číslo: 867062-95-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR