Tisk…

Biotin-PEG4-acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Biotin-PEG4-propionic acid, 15-(Biotinamido)-4,7,10,13-tetraoxapentadecanoic Acid, PEG4-biotin acid, 17-Oxo-21-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13-tetraoxa-16-azahenicosanoic acid

BIPG1199-100MG BIPG1199-250MG
APOSBIPG1199-100MGEA 6020 CZK
APOSBIPG1199-100MG APOSBIPG1199-250MG
Biotin-PEG4-acid
Biotin-PEG4-acid
Vzorec: C₂₁H₃₇N₃O₈S
MW: 491,60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08457831
CAS číslo: 721431-18-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR