Tisk…

Amino-PEG8-t-butyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 1-amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate, Amino-PEG8-Boc

BIPG1076-250MG BIPG1076-100MG BIPG1076-500MG
APOSBIPG1076-250MGEA 6780 CZK
APOSBIPG1076-250MG APOSBIPG1076-100MG APOSBIPG1076-500MG
Amino-PEG8-t-butyl ester
Amino-PEG8-t-butyl ester
Vzorec: C₂₃H₄₇NO₁₀
MW: 497,62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11041142
CAS číslo: 756526-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR