Tisk…

Amino-PEG6-t-butyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-butyl 1-amino-3,6,9,12,15,18-hexaoxahenicosan-21-oate, NH2-PEG6-Boc

BIPG1075-250MG BIPG1075-100MG BIPG1075-500MG
APOSBIPG1075-250MGEA 3540 CZK
APOSBIPG1075-250MG APOSBIPG1075-100MG APOSBIPG1075-500MG
Amino-PEG6-t-butyl ester
Amino-PEG6-t-butyl ester
Vzorec: C₁₉H₃₉NO₈
MW: 409,51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184944
CAS číslo: 1286281-32-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR