Tisk…

PIPES monosodná sůl ≥97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid) monosodium salt, Piperazine-N,N'-bis-2-ethanesulphonic acid monosodium salt, PIPES-Na, Sodium hydrogen piperazine-1,4-diethanesulphonate, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid monosodium salt, Monosodium piperazine-1,4-diethanesulphonate

BIP1705-100G BIP1705-500G
APOSBIP1705-100GEA 1660 CZK
APOSBIP1705-100G APOSBIP1705-500G
PIPES monosodná sůl ≥97%
PIPES monosodium salt

  • Buffer

Vzorec: C₈H₁₇N₂NaO₆S₂
MW: 324,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065472
CAS číslo: 10010-67-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR