Phorbol-12,13-dibutyrate ≥99%
Dodavatel: Apollo Scientific
|
Varování
|
BIP1016-5MG
BIP1016-1MG
APOSBIP1016-5MGEA
9330
CZK
APOSBIP1016-5MG
APOSBIP1016-1MG
Phorbol-12,13-dibutyrate ≥99%
Phorbol-12,13-dibutyrate
Widely used PKC activator; less potent than PMA, but also less hydrophobic, making it easier to wash out of cells in tissue culture.
Vzorec:
C₂₈H₄₀O₈ MW: 504,6 g/mol Storage Temperature: Freezer |
CAS číslo:
37558-16-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...