Tisk…

Phorbol ≥99%

Dodavatel: Apollo Scientific
Nebezpečí

BIP1010-25MG BIP1010-1MG BIP1010-10MG
APOSBIP1010-25MGEA 4960 CZK
APOSBIP1010-25MG APOSBIP1010-1MG APOSBIP1010-10MG
Phorbol ≥99%
Phorbol

Unsubstituted parent diterpene from which phorbol esters are derived; biologically inactive in most assays

Vzorec: C₂₀H₂₈O₆
Storage Temperature: Freezer
MDL Number: MFCD00065450
CAS číslo: 17673-25-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR