Penitrem A (from Penicillium paxilli)
Dodavatel: Apollo Scientific
|
Nebezpečí
|
BIP1006-10MG
BIP1006-1MG
BIP1006-5MG
APOSBIP1006-10MGEA
20470
CZK
APOSBIP1006-10MG
APOSBIP1006-1MG
APOSBIP1006-5MG
Penitrem A (from Penicillium paxilli)
Penitrem A
Tremorgenic alkaloid
Vzorec:
C₃₇H₄₄ClNO₆ Bod tání: 230 °C Storage Temperature: Freezer |
CAS číslo:
12627-35-9 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...