Tisk…

Puromycin aminonucleoside

Dodavatel: Apollo Scientific
Varování

BIP1001-100MG BIP1001-25MG BIP1001-10MG
APOSBIP1001-100MGEA 17860 CZK
APOSBIP1001-100MG APOSBIP1001-25MG APOSBIP1001-10MG
Puromycin aminonucleoside
Puromycin aminonucleoside
Vzorec: C₁₂H₁₈N₆O₃
Bod tání: 218…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063462
CAS číslo: 58-60-6
EINECS: 200-388-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR