Tisk…

PIPES disodium salt ≥98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Piperazine-N,N'-bis-2-ethanesulphonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) disodium salt, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid disodium salt, PIPES-Na2, 1,4-Piperazinediethanesulphonic acid disodium salt, Disodium piperazine-1,4-diethanesulphonate

BIP0704-100G BIP0704-500G BIP0704-1KG
APOSBIP0704-100GEA 622 CZK
APOSBIP0704-100G APOSBIP0704-500G APOSBIP0704-1KG
PIPES disodium salt ≥98%
PIPES disodium salt

  • Buffer

Vzorec: C₈H₁₆N₂Na₂O₆S₂
MW: 346.34
Storage Temperature: Ambient
MDL Number: MFCD00064349
CAS číslo: 76836-02-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR