Tisk…

Potassium-1-carboxyvinyl hydrogenphosphate 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium

BIP0691-1G BIP0691-5G BIP0691-25G
APOSBIP0691-1GEA 1330 CZK
APOSBIP0691-1G APOSBIP0691-5G APOSBIP0691-25G
Potassium-1-carboxyvinyl hydrogenphosphate 99%
Potassium-1-carboxyvinyl hydrogenphosphate
Vzorec: C₃H₄KO₆P
Storage Temperature: Freezer
MDL Number: MFCD00044476
CAS číslo: 4265-07-0
EINECS: 224-247-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR