Tisk…

Polymyxin B sulphate

Dodavatel: Apollo Scientific
Varování

BIP0145-10G BIP0145-1G BIP0145-5G
APOSBIP0145-10GEA 6220 CZK
APOSBIP0145-10G APOSBIP0145-1G APOSBIP0145-5G
Polymyxin B sulphate
Polymyxin B sulphate

Poly B is a mixture of the sulphates of polypeptides


  • approx. 8000 units/mg
  • Cell culture tested

Vzorec: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385,61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00131991
CAS číslo: 1405-20-5
EINECS: 215-774-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR