Tisk…

Penicillin V potassium salt

Dodavatel: Apollo Scientific
Nebezpečí

BIP0143-25G BIP0143-5G
APOSBIP0143-25GEA 3310 CZK
APOSBIP0143-25G APOSBIP0143-5G
Penicillin V potassium salt
Penicillin V potassium salt

Penicillin V is an inhibitor of bacterial cell wall synthesisActive against gram-positive and some gram-negative bacteria. b-lactamase sensitive.


  • Potency: 1650mg/mg

Vzorec: C₁₆H₁₇KN₂O₅S
MW: 388,48 g/mol
Bod tání: 197…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051771
CAS číslo: 132-98-9
EINECS: 205-086-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR