Tisk…

Oil red O

Dodavatel: Apollo Scientific
Varování

Synonyms: CI 26125

BIO1320-100G BIO1320-25G
APOSBIO1320-100GEA 2480 CZK
APOSBIO1320-100G APOSBIO1320-25G
Oil red O
Oil red O
Vzorec: C₂₆H₂₄N₄O
MW: 408,51 g/mol
Bod tání: 120…126 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00003898
CAS číslo: 1320-06-5
EINECS: 215-295-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR