Tisk…

Oxalacetic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid

BIO0513-100G BIO0513-25G BIO0513-50G
APOSBIO0513-100GEA 14880 CZK
APOSBIO0513-100G APOSBIO0513-25G APOSBIO0513-50G
Oxalacetic acid
Oxalacetic acid
Vzorec: C₄H₄O₅
MW: 132,07 g/mol
Bod tání: 161 °C (rozkl.)
Bod vzplanutí: 88 °C
Storage Temperature: Freezer
MDL Number: MFCD00002592
CAS číslo: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR