Tisk…

Novobiocin sodium salt ≥95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Sodium 4-((7-(((2R,3R,4S,5R)-4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-8-methyl-2-oxo-2H-chromen-3-yl)carbamoyl)-2-(3-methylbut-2-en-1-yl)phenolate, Novobiocin sodium

BIN0394-25G BIN0394-10G BIN0394-5G
APOSBIN0394-25GEA 10750 CZK
APOSBIN0394-25G APOSBIN0394-10G APOSBIN0394-5G
Novobiocin sodium salt ≥95%
Novobiocin sodium salt
Vzorec: C₃₁H₃₅N₂NaO₁₁
MW: 634,614 g/mol
Bod tání: 215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066541
CAS číslo: 1476-53-5
EINECS: 216-023-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR