Tisk…

Nitrocefin

Dodavatel: Apollo Scientific
Varování

Synonyms: (6R,7R)-7-[[(E)-4-(2,4-Dinitrophenyl)-2-(2-thienyl)but-3-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-(2,4-DINITROSTYRYL)-(6R, 7R)-7-(2-THIENYLACETAMIDO)-CEPH-3-EM-4 CARBOXYLIC ACID, E-ISOMER, N-[3-[2-(2,4-DINITRO-PHENYL)-VINYL]-8-OXO-5-THIA-1-AZA-BICYCLO[4.2.0]OCT-7-YL]-2-THIOPHEN-2-YL-ACETAMIDE

BIN0140-10MG BIN0140-1MG BIN0140-5MG
APOSBIN0140-10MGEA 11580 CZK
APOSBIN0140-10MG APOSBIN0140-1MG APOSBIN0140-5MG
Nitrocefin
Nitrocefin
Vzorec: C₂₁H₁₆N₄O₈S₂
MW: 516,499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00864896
CAS číslo: 41906-86-9
UN: 1325
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR