Tisk…

Nitrofurantoin

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-[(5-Nitrofurfurylidene)amino]hydantoin

BIN0136-100G BIN0136-250G BIN0136-500G
APOSBIN0136-100GEA 1700 CZK
APOSBIN0136-100G APOSBIN0136-250G APOSBIN0136-500G
Nitrofurantoin
Nitrofurantoin

Nitrofurantoin is bactericidal in vitro to most gram-positive and gram-negative bacteria.

Vzorec: C₈H₆N₄O₅
MW: 238,16 g/mol
Bod tání: 267…269 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00003224
CAS číslo: 67-20-9
EINECS: 200-646-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR