Tisk…

Nonactin (from Streptomyces griseus)

Dodavatel: Apollo Scientific
Varování

Synonyms: Polynactin, Werramycin A

BIN0110-10MG BIN0110-25MG
APOSBIN0110-10MGEA 2900 CZK
APOSBIN0110-10MG APOSBIN0110-25MG
Nonactin (from Streptomyces griseus)
Nonactin
Vzorec: C₄₀H₆₄O₁₂
MW: 736.9 g/mol
Storage Temperature: Freezer
CAS číslo: 6833-84-7
EINECS: 229-911-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR