Tisk…

3'-Amino-2',3'-dideoxyadenosine

Dodavatel: Apollo Scientific

BIMN2002-100MG BIMN2002-1G BIMN2002-250MG
APOSBIMN2002-100MGEA 1210 CZK
APOSBIMN2002-100MG APOSBIMN2002-1G APOSBIMN2002-250MG
3'-Amino-2',3'-dideoxyadenosine
3'-Amino-2',3'-dideoxyadenosine
Vzorec: C₁₀H₁₄N₆O₂
MW: 250,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703999
CAS číslo: 7403-25-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR