Tisk…

Genistein ≥98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

BIMB2006-1G BIMB2006-500MG
APOSBIMB2006-1GEA 6190 CZK
APOSBIMB2006-1G APOSBIMB2006-500MG
Genistein ≥98%
Genistein

Inhibitor of tyrosine protein kinase.

Vzorec: C₁₅H₁₀O₅
MW: 270,24 g/mol
Bod tání: 297…298 °C
Storage Temperature: Freezer
MDL Number: MFCD00016952
CAS číslo: 446-72-0
EINECS: 207-174-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR