Tisk…
5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98%
Dodavatel: Apollo Scientific
Varování
Synonyms: Magenta-Gal, Mag-Gal
BIMB1331-1G BIMB1331-2.5G BIMB1331-250MG
APOSBIMB1331-1GEA 2110 CZK
APOSBIMB1331-1G APOSBIMB1331-2.5G APOSBIMB1331-250MG
5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98%
5-Bromo-6-chloro-3-indolyl-beta-D-galactopyranoside
Magenta Gal is a chromogenic substrate forb-Galactosidase, producing a red or magenta insoluble product.
Vzorec: C₁₄H₁₅BrClNO₆
MW: 408,63 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210022
CAS číslo: 93863-88-8