Tisk…

5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Magenta-GlcA CHA Salt, Magenta-Gluc CHA Salt, 5-Bromo-6-chloro-3-indolyl-β-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt

BIMB1302-1G BIMB1302-250MG BIMB1302-500MG
APOSBIMB1302-1GEA 6620 CZK
APOSBIMB1302-1G APOSBIMB1302-250MG APOSBIMB1302-500MG
5-Bromo-6-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥98%
5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt

Alternative to 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide(X-GlcA) for detection of b-Glucuronidase gene.

Vzorec: C₁₄H₁₃BrClNO₇
MW: 521,79 g/mol
Bod tání: 216…217 °C
Storage Temperature: Freezer
MDL Number: MFCD00153929
CAS číslo: 144110-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR