Tisk…
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
Dodavatel: Apollo Scientific
Varování
Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
BIMB1018-1G BIMB1018-250MG BIMB1018-5G
APOSBIMB1018-1GEA 2360 CZK
APOSBIMB1018-1G APOSBIMB1018-250MG APOSBIMB1018-5G
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Substrate for Alkaline phosphatase.
  • Soluble in water
Vzorec: C₈H₄BrClNNa₂O₄P
MW: 370,43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036757
CAS číslo: 102185-33-1