Tisk…
Nω-Nitro-L-arginine methyl ester hydrochloride
Dodavatel: Apollo Scientific
Synonyms: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride
BIMB1017-5G BIMB1017-25G
APOSBIMB1017-5GEA 2360 CZK
APOSBIMB1017-5G APOSBIMB1017-25G
Nω-Nitro-L-arginine methyl ester hydrochloride
Nomega-Nitro-L-arginine methyl ester hydrochloride
Vzorec: C₇H₁₅N₅O₄
MW: 269.69 g/mol
Bod tání: 159…161 °C (rozkl.)
Storage Temperature: Freezer
MDL Number: MFCD00039052
CAS číslo: 51298-62-5