4-MUG ≥98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid, 4-methylumbelliferyl-beta-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronide
BIMB1003-1G
BIMB1003-5G
BIMB1003-10G
APOSBIMB1003-1GEA
1040
CZK
APOSBIMB1003-1G
APOSBIMB1003-5G
APOSBIMB1003-10G
4-MUG ≥98%
4-MUG
Fluorescent substrate for b-D-glucuronidase (GUS) encoded by the gusA gene isolated originally from E.Coli.
- Soluble in DMFO and DMSO
Vzorec:
C₁₆H₁₆O₉ MW: 352,3 g/mol Bod tání: 178…180 °C Storage Temperature: Freezer |
MDL Number:
MFCD00150472 CAS číslo: 6160-80-1 EINECS: 228-186-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...