Tisk…

(3-Indoxyl)-β-D-glucopyranoside

Dodavatel: Apollo Scientific
Varování

Synonyms: Indoxyl-D-glucoside, 3-Indole-D-glucoside, 3-Indolyl-D-glucoside, Indoxyl-B-D Glucoside, Indican (Plant Indican)

BIMB0907-1G BIMB0907-250MG BIMB0907-500MG
APOSBIMB0907-1GEA 4370 CZK
APOSBIMB0907-1G APOSBIMB0907-250MG APOSBIMB0907-500MG
(3-Indoxyl)-β-D-glucopyranoside
(3-Indoxyl)-β-D-glucopyranoside
Vzorec: C₁₄H₁₇NO₆
Bod tání: 57…58 °C
Storage Temperature: Freezer
MDL Number: MFCD00047169
CAS číslo: 487-60-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR