Tisk…

Methyl-1,2,3,4-tetra-O-acetyl-β-D-glucuronate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronate, (2S,3R,4S,5S,6S)-6-(Methoxycarbonyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4-Tetra-O-acetyl-β-D-glucuronic acid Methyl Ester, 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronic acid Methyl Ester, Methyl tetra-O-acetyl-β-D-glucopyranuronate

BIM6000-1G BIM6000-250MG BIM6000-5G
APOSBIM6000-1GEA 1580 CZK
APOSBIM6000-1G APOSBIM6000-250MG APOSBIM6000-5G
Methyl-1,2,3,4-tetra-O-acetyl-β-D-glucuronate
Methyl-1,2,3,4-tetra-O-acetyl-beta-D-glucuronate
Vzorec: C₁₅H₂₀O₁₁
MW: 376,31 g/mol
Bod tání: 177…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069834
CAS číslo: 7355-18-2
EINECS: 230-880-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR