Tisk…

Lutein

Dodavatel: Apollo Scientific

Synonyms: E 161b, (6'R)-β,ε-carotene-3R,3'R-diol, Xanthophyll, (1R,4R)-4-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-((R)-4-Hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl)-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl)-3,5,5-trimethylcyclohex-2-enol

BIM1909-5MG BIM1909-10MG BIM1909-25MG
APOSBIM1909-5MGEA 2690 CZK
APOSBIM1909-5MG APOSBIM1909-10MG APOSBIM1909-25MG
Lutein
Lutein
Vzorec: C₄₀H₅₆O₂
MW: 568,88 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08435941
CAS číslo: 127-40-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR