Tisk…

2-Nitrophenyl-β-D-glucoside

Dodavatel: Apollo Scientific
Varování

BIM1206-5G
APOSBIM1206-5GEA 6620 CZK
APOSBIM1206-5G
2-Nitrophenyl-β-D-glucoside
2-Nitrophenyl-β-D-glucoside

Substrate for b-D-Glucosidase

Vzorec: C₁₂H₁₅NO₈
MW: 301,25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065048
CAS číslo: 2816-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR