4-Nitrophenyl-ɑ-L-fucopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
BIM1205-1G
BIM1205-250MG
APOSBIM1205-1GEA
12200
CZK
APOSBIM1205-1G
APOSBIM1205-250MG
4-Nitrophenyl-ɑ-L-fucopyranoside
4-Nitrophenyl-alpha-L-fucopyranoside
Vzorec:
C₁₂H₁₅NO₇ MW: 285,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063697 CAS číslo: 10231-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...