Tisk…

4-Nitrophenyl-N-acetyl-b-D-galactosaminide

Dodavatel: Apollo Scientific
Varování

Synonyms: Gb5-β-pNP, 4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside, 4-Nitrophenyl N-Acetyl-beta-D-galactosaminide, N-((2S,3R,4R,5R,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)tetrahydro-2H-pyran-3-yl)acetamide

BIM1204-1G BIM1204-250MG BIM1204-500MG
APOSBIM1204-1GEA 6220 CZK
APOSBIM1204-1G APOSBIM1204-250MG APOSBIM1204-500MG
4-Nitrophenyl-N-acetyl-b-D-galactosaminide
4-Nitrophenyl-N-acetyl-b-D-galactosaminide

Substrate for N-Acetyl-b-D-glucosaminidase

Vzorec: C₁₄H₁₈N₂O₈
MW: 342,31 g/mol
Bod tání: 207 °C
Storage Temperature: Freezer
MDL Number: MFCD00067360
CAS číslo: 14948-96-0
EINECS: 239-024-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR