4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
GLCNAC1-B-PNP, 4-Nitrophenyl N-Acetyl-β-D-glucosaminide, 4-Nitrophenyl 2-acetamido-2-deoxy-beta-D-glucopyranoside
BIM1201-1G
BIM1201-250MG
APOSBIM1201-1GEA
2070
CZK
APOSBIM1201-1G
APOSBIM1201-250MG
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside
4-Nitrophenyl-2-acetamido-2-deoxy-beta-D-glucopyranoside
Substrate for N-Acetyl-b-D-glucosaminidase
Vzorec:
C₁₄H₁₈N₂O₈ MW: 342.30 g/mol Bod tání: 210…212 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063696 CAS číslo: 3459-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...