Tisk…

Ponatinib free base 99+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(2-Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-benzamide, 3-(Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide, AP24534

BIM1122-100MG
APOSBIM1122-100MGEA 3110 CZK
APOSBIM1122-100MG
Ponatinib free base 99+%
Ponatinib
Vzorec: C₂₉H₂₇F₃N₆O
MW: 532,56 g/mol
Bod tání: 190 °C
Storage Temperature: Freezer
MDL Number: MFCD17215203
CAS číslo: 943319-70-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR