4-Methylumbelliferyl-β-D-xylopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Methyl-7-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate
BIM1103-1G
BIM1103-250MG
APOSBIM1103-1GEA
4960
CZK
APOSBIM1103-1G
APOSBIM1103-250MG
4-Methylumbelliferyl-β-D-xylopyranoside
4-Methylumbelliferyl-beta-D-xylopyranoside
Vzorec:
C₁₅H₁₆O₇ MW: 308,286 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00037607 CAS číslo: 6734-33-4 EINECS: 229-784-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...