Tisk…

N-Acetyl-D-galactosamine ≥98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

BIM1065-1G BIM1065-5G
APOSBIM1065-1GEA 2280 CZK
APOSBIM1065-1G APOSBIM1065-5G
N-Acetyl-D-galactosamine ≥98%
N-Acetyl-D-galactosamine
Vzorec: C₈H₁₅NO₆
MW: 221,21 g/mol
Bod tání: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065372
CAS číslo: 1811-31-0
EINECS: 217-321-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR