Tisk…

Maltohexaose

Dodavatel: Apollo Scientific
Varování

Synonyms: (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

BIM1013-100MG BIM1013-250MG BIM1013-500MG
APOSBIM1013-100MGEA 3190 CZK
APOSBIM1013-100MG APOSBIM1013-250MG APOSBIM1013-500MG
Maltohexaose
Maltohexaose
Vzorec: C₃₆H₆₂O₃₁
MW: 990,8589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083650
CAS číslo: 34620-77-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR