Tisk…

(4-Chloro-2-methylphenoxy)acetic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: MCPA

BIM1010-25G BIM1010-100G
APOSBIM1010-25GEA 827 CZK
APOSBIM1010-25G APOSBIM1010-100G
(4-Chloro-2-methylphenoxy)acetic acid 98%
(4-Chloro-2-methylphenoxy)acetic acid
Vzorec: C₉H₉ClO₃
MW: 200,62 g/mol
Bod tání: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004306
CAS číslo: 94-74-6
UN: 3077
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR