Methotrexate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
BIM0130-1G
BIM0130-5G
BIM0130-10G
APOSBIM0130-1GEA
622
CZK
APOSBIM0130-1G
APOSBIM0130-5G
APOSBIM0130-10G
Methotrexate
Methotrexate
An antineoplastic agent which acts as an antimetaboliteof folic acid.
Vzorec:
C₂₀H₂₂N₈O₅ MW: 454,44 g/mol Bod tání: 185…204 °C Storage Temperature: Freezer |
MDL Number:
MFCD00150847 CAS číslo: 59-05-2 UN: 1544 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...