Cefoxitin sodium
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
BIM0108-1G
BIM0108-250MG
BIM0108-5G
APOSBIM0108-1GEA
1870
CZK
APOSBIM0108-1G
APOSBIM0108-250MG
APOSBIM0108-5G
Cefoxitin sodium
Cefoxitin sodium
Vzorec:
C₁₆H₁₆N₃NaO₇S₂ MW: 449.428 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00079042 CAS číslo: 33564-30-6 EINECS: 251-574-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...