Tisk…

Oxacillin sodium salt monohydrate

Dodavatel: Apollo Scientific
Varování

BIM0106-1G BIM0106-25G BIM0106-5G
APOSBIM0106-1GEA 622 CZK
APOSBIM0106-1G APOSBIM0106-25G APOSBIM0106-5G
Oxacillin sodium salt monohydrate
Oxacillin sodium salt monohydrate
Vzorec: C₁₉H₁₉N₃O₅S
MW: 423,42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00167146
CAS číslo: 7240-38-2
EINECS: 214-636-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR