Tisk…
D(-)-Luciferin potassium salt
Dodavatel: Apollo Scientific
Varování
Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
BIL1800-100MG BIL1800-250MG BIL1800-50MG
APOSBIL1800-100MGEA 1210 CZK
APOSBIL1800-100MG APOSBIL1800-250MG APOSBIL1800-50MG
D(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
Vzorec: C₁₁H₇KN₂O₃S₂
MW: 318.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00044928
CAS číslo: 115144-35-9