Tisk…

D-(-)-Luciferin free acid ≥99.5%

Dodavatel: Apollo Scientific
Varování

Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

BIL1349-100MG BIL1349-250MG BIL1349-50MG
APOSBIL1349-100MGEA 1910 CZK
APOSBIL1349-100MG APOSBIL1349-250MG APOSBIL1349-50MG
D-(-)-Luciferin free acid ≥99.5%
D-(-)-Luciferin

L-Luciferin: <0.5%Used with firefly luciferase (BIF1350) for the determination of ATP using bioluminescence

Vzorec: C₁₁H₈N₂O₃S₂
MW: 280,3228 g/mol
Bod tání: 200…201 °C
Storage Temperature: Freezer
MDL Number: MFCD00042929
CAS číslo: 2591-17-5
EINECS: 219-981-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR